missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cofilin 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | Cofilin 2 |
---|---|
Konzentration | 0.15 mg/mL |
Anwendungen | Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16839193
|
Proteintech
11848-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 06-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16829193
|
Proteintech
11848-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 06-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Cofilin is a widely distributed intracellular actin-modulating protein that binds and depolymerizes filamentous F-actin and inhibits the polymerization of monomeric G-actin in a pH-dependent manner. It is involved in the translocation of actin-cofilin complex from cytoplasm to nucleus.Spezifikation
Cofilin 2 | |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
P45591, Q9Y281 | |
1073, 12632, 366624 | |
Cofilin 2 Fusion Protein Ag2408 | |
Primary | |
-20°C | |
CFL2 |
0.15 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CFL2, Cofilin 2, cofilin 2 (muscle), Cofilin, muscle isoform, NEM7 | |
Cfl2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |