missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clostridium Difficile Toxin A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
239.00 CHF - 498.00 CHF
Spezifikation
| Antigen | Clostridium Difficile Toxin A |
|---|---|
| Konzentration | 5.0 mg/mL |
| Verdünnung | ELISA 1:100-1:2000 |
| Anwendungen | ELISA |
| Klassifikation | Polyclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18001416
|
Novus Biologicals
NB100-62473-0.1ML |
0.1 mL |
239.00 CHF
0.10 Milliliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18025355
|
Novus Biologicals
NB100-62473 |
0.5 mL |
498.00 CHF
0.50 Milliliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Clostridium Difficile Toxin A Polyclonal specifically detects Clostridium Difficile Toxin A in Bacteria samples. It is validated for Western Blot, ELISA.Spezifikation
| Clostridium Difficile Toxin A | |
| ELISA 1:100-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 0.09% Sodium Azide | |
| tcdA, toxA | |
| IgG | |
| Protein G purified | |
| NB100-62473 is specific for Clostridium difficile Toxin A, a gram-positive motile bacterium which is the leading cause of diarrhoea in developed countries. The incidence of disease in humans varies greatly with age, spore density, and the administration of antibiotics. Toxin A (TcdA) is a large exotoxin which is an essential virulence factor for C. difficile infection. TcdA has a molecular weight of 308kD and is a glucosyltransferase, which irreversibly inactivates the Ras family of small GTPases to disrupt many important signalling pathways. |
| 5.0 mg/mL | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Bacteria | |
| P16154 | |
| Synthetic peptide corresponding to the amino acids 2123 - 2132 (CTIDGKKYYFN) of the native molecule conjugated to Keyhole limpet Haemocyanin. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts