missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clostridium Difficile Toxin A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NB100-62473
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Clostridium Difficile Toxin A Polyclonal specifically detects Clostridium Difficile Toxin A in Bacteria samples. It is validated for Western Blot, ELISA.
Spezifikation
| Clostridium Difficile Toxin A | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.09% Sodium Azide | |
| tcdA, toxA | |
| Synthetic peptide corresponding to the amino acids 2123 - 2132 (CTIDGKKYYFN) of the native molecule conjugated to Keyhole limpet Haemocyanin. | |
| 0.5 mL | |
| Primary | |
| Bacteria | |
| Purified |
| ELISA | |
| 5.0 mg/mL | |
| ELISA 1:100-1:2000 | |
| P16154 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| NB100-62473 is specific for Clostridium difficile Toxin A, a gram-positive motile bacterium which is the leading cause of diarrhoea in developed countries. The incidence of disease in humans varies greatly with age, spore density, and the administration of antibiotics. Toxin A (TcdA) is a large exotoxin which is an essential virulence factor for C. difficile infection. TcdA has a molecular weight of 308kD and is a glucosyltransferase, which irreversibly inactivates the Ras family of small GTPases to disrupt many important signalling pathways. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction