Learn More
Invitrogen™ Human CLK2, GST Tag Recombinant Protein
CLK2 is a serine/threonine kinase that phosphorylates serine/arginine-rich proteins as related to controlling RNA splicing.
Marke: Invitrogen™ PV4201
Additional Details : Gewicht : 0.05250kg
Beschreibung
CLK2 is a serine⁄threonine kinase that phosphorylates serine⁄arginine-rich proteins as related to controlling RNA splicing. It is also thought to link cell cycle progression, apoptosis, and telomere length regulation.Spezifikation
NP_003983 | |
1196, 4776 | |
10 μg | |
CLK Kinase Family | |
Human | |
GST Tag | |
Biochemical binding/kinetic, Biochemical phospho-specific activity (no antibody) | |
Dry Ice | |
CLK2 | |
Baculovirus | |
Recombinant, Catalytic | |
Serine/Threonine Kinases |
Kinase Assay | |
70.3 kDa | |
CLK2 | |
aa 137-498 | |
Recombinant | |
Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% Glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: CMCG Group | |
Z'-LYTE™ | |
CDC-like kinase 2 | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |