Learn More
Invitrogen™ Human CLK2, GST Tag Recombinant Protein
CLK2 is a serine/threonine kinase that phosphorylates serine/arginine-rich proteins as related to controlling RNA splicing.
Marke: Invitrogen™ PV4279
Additional Details : Gewicht : 0.00900kg
Beschreibung
CLK2 is thought to link cell cycle progression, apoptosis, and telomere length regulation.
Spezifikation
NP_003983 | |
1mg | |
CLK Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% Glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Catalytic | |
Drug Development |
70.3 kDa | |
CLK2 | |
Human | |
GST Tag | |
Ser⁄Thr Kinases: CMCG Group | |
Baculovirus | |
Frozen |