missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP72 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 19928-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Involved in the recruitment of key centrosomal proteins to the centrosome. Provides centrosomal microtubule-nucleation activity on the gamma-tubulin ring complexes (gamma-TuRCs) and has critical roles in forming a focused bipolar spindle, which is needed for proper tension generation between sister chromatids. Required for localization of KIZ, AKAP9 and gamma-tubulin ring complexes (gamma-TuRCs).Spezifikation
CEP72 | |
Polyclonal | |
Unconjugated | |
CEP72 | |
centrosomal protein 72kDa, Centrosomal protein of 72 kDa, CEP72, KIAA1519 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55722, 74470 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9D3R3, Q9P209 | |
CEP72 | |
CEP72 Fusion Protein Ag13905 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |