Learn More
Centrophenoxine hydrochloride, 98%, Thermo Scientific™
41.25 CHF - 296.50 CHF
Chemische Identifikatoren
| CAS | 3685-84-5 |
|---|---|
| Summenformel | C12H17Cl2NO3 |
| Molekulargewicht (g/mol) | 294.17 |
| MDL-Nummer | MFCD00012533 |
| InChI-Schlüssel | FIVHOHCAXWQPGC-UHFFFAOYSA-N |
| Synonym | meclofenoxate hydrochloride, centrophenoxine hydrochloride, lucidril, cerutil, meclofenoxate hcl, atsefen, helfergin, marucotol, acefen, brenal |
| PubChem CID | 19379 |
| IUPAC-Name | hydrogen 2-(dimethylamino)ethyl 2-(4-chlorophenoxy)acetate chloride |
| SMILES | [H+].[Cl-].CN(C)CCOC(=O)COC1=CC=C(Cl)C=C1 |
| Product Code | Brand | Menge | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Menge | Price | Quantity & Availability | |||||
|
15466259
|
Thermo Scientific Alfa Aesar
J64196.06 |
5g |
41.25 CHF
5 Gramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
15476259
|
Thermo Scientific Alfa Aesar
J64196.14 |
25g |
86.52 CHF
25 Gramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
15486259
|
Thermo Scientific Alfa Aesar
J64196.22 |
100g |
296.50 CHF
100 Gramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Ester of dimethylethanolamine and 4-chlorophenoxyacetic acid; activates acetylcholinesterase activity in hippocampus of aged rats
Chemical Identifiers
| C12H17Cl2NO3 | |
| MFCD00012533 | |
| meclofenoxate hydrochloride, centrophenoxine hydrochloride, lucidril, cerutil, meclofenoxate hcl, atsefen, helfergin, marucotol, acefen, brenal | |
| hydrogen 2-(dimethylamino)ethyl 2-(4-chlorophenoxy)acetate chloride |
Specifications
| Centrophenoxine hydrochloride | |
| C12H17Cl2NO3 | |
| 14,5780 | |
| FIVHOHCAXWQPGC-UHFFFAOYSA-N | |
| hydrogen 2-(dimethylamino)ethyl 2-(4-chlorophenoxy)acetate chloride | |
| 19379 | |
| 98% |
| 3685-84-5 | |
| MFCD00012533 | |
| meclofenoxate hydrochloride, centrophenoxine hydrochloride, lucidril, cerutil, meclofenoxate hcl, atsefen, helfergin, marucotol, acefen, brenal | |
| [H+].[Cl-].CN(C)CCOC(=O)COC1=CC=C(Cl)C=C1 | |
| 294.17 | |
| 294.17 |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'einecsNumber' : 222-975-3
missing translation for 'rtecsNumber' : AG0440000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.