Learn More
Greiner Bio-One CELLSTAR Cell-Repellent Surface Cell Culture Flasks with Filter Screw Cap
Ideal for suspension, spheroid cultures and stem cell aggregate formation. Greiner Bio-One CELLSTAR Cell-Repellent Surface Cell Culture Flasks with Filter Screw Cap effectively inhibit cell adhesion.
Marca:  Greiner Bio-One 658985
Detalles adicionales : Gewicht : 0.40000kg
 Ver más versiones de este producto
Código de producto. 17464532
  • 936.00 CHF / 1 Stück
Verfügbar ab: 20-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Añadir a la cesta
Descripción
Descripción
  • Made of polystyrene
  • Used for applications such as Gel-based 3D cultures and Magnetic 3D cell culturing (Nano3D technology)
  • Cell-repellent surface does not degrade or leach under common cell culture conditions, rendering an ideal substrate for native cell culture experiments
Especificaciones
Especificaciones
White
Cell Repellent
250 mL
Polystyrene
Filter Screw Cap
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificats