Learn More
Greiner Bio-One CELLSTAR Cell-Repellent Surface Cell Culture Flasks with Filter Screw Cap
Ideal for suspension, spheroid cultures and stem cell aggregate formation. Greiner Bio-One CELLSTAR Cell-Repellent Surface Cell Culture Flasks with Filter Screw Cap effectively inhibit cell adhesion.
Marca: Greiner Bio-One 658985
Detalles adicionales : Gewicht : 0.40000kg
Descripción
- Made of polystyrene
- Used for applications such as Gel-based 3D cultures and Magnetic 3D cell culturing (Nano3D technology)
- Cell-repellent surface does not degrade or leach under common cell culture conditions, rendering an ideal substrate for native cell culture experiments
Especificaciones
White | |
Cell Repellent | |
250 mL |
Polystyrene | |
Filter Screw Cap |