Learn More
CD81 Mouse anti-Human, Unconjugated, Clone: 1G2C6, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66866-1-IG-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The CD81 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD81. This antibody recognizes human antigen. The CD81 antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Spezifikation
| CD81 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry, ELISA | |
| 1G2C6 | |
| Unconjugated | |
| 20 μL | |
| 975 | |
| IgG1 |