missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD57 Mouse anti-Human, Clone: 1F4A11, Proteintech
Mouse Monoclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | CD57 |
---|---|
Klon | 1F4A11 |
Konzentration | 1.13 mg/mL |
Anwendungen | Western Blot |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16848845
|
Proteintech
66711-1-IG-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16838845
|
Proteintech
66711-1-IG-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Natural killer (NK) cells in teleosts and their evolutionary homologue are a subpopulation of lymphocytes with properties that distinguish them from either B- or T-cells. NK cells are important effectors of innate immunity where they release cytokines, which in turn up-regulate other immunological functions. Monoclonal antibodies have been used to identify different surface antigens present on NK cells. These surface antigens have not only been used to identify NK cells, but also their functionally distinct subsets. The Cluster of Differentiation (CD) nomenclature was established to standardize the naming of NK cells.Spezifikation
CD57 | |
1.13 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
B3GAT1, CD57, GlcAT P, GLCATP, GlcUAT P, Glucuronosyltransferase P, HNK 1, HNK1, LEU7, NK 1 | |
B3GAT1 | |
IgG2a | |
Protein A | |
Antibody |
1F4A11 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q9P2W7 | |
27087 | |
CD57 Fusion Protein Ag13593 | |
Primary | |
-20°C | |
B3GAT1 |