missing translation for 'onlineSavingsMsg'
Learn More

CD57 Mouse anti-Human, Clone: 1F4A11, Proteintech

Mouse Monoclonal Antibody

Marke:  Proteintech 66711-1-IG-20UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16848845

  • 133.80 CHF / 20 Mikroliter
Verfügbar ab: 24-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Natural killer (NK) cells in teleosts and their evolutionary homologue are a subpopulation of lymphocytes with properties that distinguish them from either B- or T-cells. NK cells are important effectors of innate immunity where they release cytokines, which in turn up-regulate other immunological functions. Monoclonal antibodies have been used to identify different surface antigens present on NK cells. These surface antigens have not only been used to identify NK cells, but also their functionally distinct subsets. The Cluster of Differentiation (CD) nomenclature was established to standardize the naming of NK cells.
Spezifikation

Spezifikation

CD57
Monoclonal
1.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9P2W7
B3GAT1
CD57 Fusion Protein Ag13593
20 μL
Primary
Human
Antibody
IgG2a
Western Blot
1F4A11
Unconjugated
B3GAT1
B3GAT1, CD57, GlcAT P, GLCATP, GlcUAT P, Glucuronosyltransferase P, HNK 1, HNK1, LEU7, NK 1
Mouse
Protein A
RUO
27087
-20°C
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote