missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45 Rat, Biotin, Clone: 30-F11, enQuireBio™
Rat Monoclonal Antibody
Marke: EnquireBio QAB40-B-100ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
CD45 | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH7.2 | |
Ptprc | |
Protein A/G Purified | |
RUO | |
19264 | |
IgG2b κ |
Flow Cytometry | |
30-F11 | |
Biotin | |
NP_001255215 | |
Rat | |
100 μg | |
Primary | |
Store at 2-4°C. |