missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD31 / PECAM-1 Rabbit, Clone: C31/1395R, enQuireBio™
Rabbit Monoclonal Antibody
Marke: EnquireBio 5175-RBM11-P1
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
| CD31 / PECAM-1 | |
| Cell surface and cytoplasmic | |
| C31/1395R | |
| Unconjugated | |
| XP_005276940 | |
| Rabbit | |
| ∽100kDa (endothelium) and ∽130kDa (platelets) | |
| Protein A/G Purified | |
| RUO | |
| 5175 | |
| IgG κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with or without 0.05% BSA and 0.05% azide. | |
| PECAM1 | |
| Human recombinant full-length CD31 protein | |
| Jurkat cells. Tonsil or Angiosarcoma. | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Berichtigung von Produktinhalten
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction