missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD252 / OX40-L Rat, APC, Clone: RM134L, enQuireBio™
Rat Monoclonal Antibody
Marke: EnquireBio QAB75-APC-100ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
CD252 / OX40-L | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
Tnfsf4 | |
Protein A/G Purified | |
RUO | |
22164 | |
IgG2b κ |
Flow Cytometry | |
RM134L | |
APC | |
NP_033478 | |
Rat | |
100 μg | |
Primary | |
Store at 2-4°C. |