missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD24 Rat, PE, Clone: M1/69, enQuireBio™
Rat Monoclonal Antibody
Marke: EnquireBio QAB33-PE-100ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
CD24 | |
Monoclonal | |
0.2mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Cd24a | |
Protein A/G Purified | |
RUO | |
12484 | |
IgG2b κ |
Flow Cytometry | |
M1/69 | |
PE | |
NP_033976 | |
Rat | |
100 μg | |
Primary | |
Store at 2-4°C. |