Learn More
CD19 Monoclonal Antibody (eBio1D3 (1D3)), Functional Grade, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
3280.00 CHF - 16350.00 CHF
Spezifikation
| Antigen | CD19 |
|---|---|
| Klon | eBio1D3 (1D3) |
| Konzentration | 1 mg/mL |
| Anwendungen | Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen) |
| Klassifikation | Monoclonal |
Beschreibung
The eBio1D3 MAb reacts with mouse CD19, a 95 kDa transmembrane glycoprotein. CD19 is expressed by B cells during all stages of development excluding the terminally differentiated plasma cells. Follicular dendritic cells also express CD19. Together CD21, CD81, MHC class II, and CD19 form a multimolecular complex that associates with the BCR. Signaling through CD19 induces tyrosine phosphorylation, calcium flux and proliferation of B cells.
CD19 Monoclonal antibody specifically detects CD19 in Mouse samples. It is validated for Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen)Spezifikation
| CD19 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.2 | |
| AW495831; B4; B-lymphocyte antigen CD19; B-lymphocyte surface antigen B4; Cd19; CD19 antigen; CD19 molecule; CVID3; differentiation antigen CD19; Leu-12; T-cell surface antigen Leu-12 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
| eBio1D3 (1D3) | |
| Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen) | |
| Functional Grade | |
| Rat | |
| Mouse | |
| P25918 | |
| 12478 | |
| Primary | |
| 4° C | |
| Cd19 |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.