missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD161 Mouse, PE, Clone: HP-3G10, enQuireBio™
Mouse Monoclonal Antibody
Marke: EnquireBio QAB60-PE-100Tests
Additional Details : Gewicht : 0.01000kg
Spezifikation
CD161 | |
Monoclonal | |
5μL/Test | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
KLRB1 | |
Protein A/G Purified | |
RUO | |
3820 | |
IgG1 κ |
Flow Cytometry | |
HP-3G10 | |
PE | |
NP_002249 | |
Mouse | |
100 Tests | |
Primary | |
Store at 2-4°C. |