missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD152 / CTLA-4 Armenian Hamster, PE-Cy7, Clone: UC10-4F10-11, enQuireBio™
Armenian Hamster Monoclonal Antibody
Marke: EnquireBio QAB58-PE7-100ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
CD152 / CTLA-4 | |
Monoclonal | |
0.2mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Ctla4 | |
Protein A/G Purified | |
RUO | |
12477 | |
IgG |
Flow Cytometry | |
UC10-4F10-11 | |
PE-Cyanine7 | |
NP_001268905 | |
Armenian Hamster | |
100 μg | |
Primary | |
Store at 2-4°C. |