missing translation for 'onlineSavingsMsg'
Learn More

CD13 Mouse anti-Human, APC, Clone: WM15, BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 561698

Additional Details : Gewicht : 0.02000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15831249

  • 235.00 CHF / 1 Stück
Verfügbar ab: 16-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The WM15 monoclonal antibody specifically binds to the 150 kDa Type II integral membrane glycoprotein aminopeptidase N. This antibody binds to GM-progenitor cells, granulocytic and monocytic cells, and mast cells, but not to lymphocytes, platelets or erythrocytes. Aminopeptidase N is involved in the metabolism of many regulatory peptides.

Spezifikation

Spezifikation

CD13
Monoclonal
APC
Mouse
Affinity Purified
RUO
Human
IgG1 κ
Flow Cytometry
WM15
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human AML Cells
25 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only.