missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCT4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21524-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The chaperonin containing TCP1 complex (CCT), also called the TCP1 ring complex, consists of 2 back-to-back rings, each containing 8 unique but homologous subunits, such as CCT4. CCT assists the folding of newly translated polypeptide substrates through multiple rounds of ATP-driven release and rebinding of partially folded intermediate forms. Substrates of CCT include the cytoskeletal proteins actin and tubulin, as well as alpha-transducin.Spezifikation
CCT4 | |
Polyclonal | |
Unconjugated | |
CCT4 | |
CCT DELTA, CCT4, Cctd, SRB, Stimulator of TAR RNA binding, TCP 1 delta | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10575, 12464, 29374 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P50991, P80315, Q7TPB1 | |
CCT4 | |
CCT4 Fusion Protein Ag16128 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |