missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCNDBP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12363-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene was identified by the interaction of its gene product with Grap2, a leukocyte-specific adaptor protein important for immune cell signaling. The protein encoded by this gene was shown to interact with cyclin D. Transfection of this gene in cells was reported to reduce the phosphorylation of Rb gene product by cyclin D-dependent protein kinase, and inhibit E2F1-mediated transcription activity. This protein was also found to interact with helix-loop-helix protein E12 and is thought to be a negative regulator of liver-specific gene expression. Two alternatively spliced variants, which encode distinct isoforms, have been reported.Spezifikation
CCNDBP1 | |
Polyclonal | |
Unconjugated | |
CCNDBP1 | |
CCNDBP1, Cyclin D1 binding protein 1, DIP1, GCIP, HHM, Human homolog of Maid | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17151, 23582, 362201 | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O95273, Q3TVC7, Q5BK06 | |
Ccndbp1 | |
CCNDBP1 Fusion Protein Ag3032 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |