missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCNDBP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | CCNDBP1 |
---|---|
Konzentration | 0.28 mg/mL |
Anwendungen | Western Blot, Immunohistochemistry (Paraffin) |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16850044
|
Proteintech
12363-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16840044
|
Proteintech
12363-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
This gene was identified by the interaction of its gene product with Grap2, a leukocyte-specific adaptor protein important for immune cell signaling. The protein encoded by this gene was shown to interact with cyclin D. Transfection of this gene in cells was reported to reduce the phosphorylation of Rb gene product by cyclin D-dependent protein kinase, and inhibit E2F1-mediated transcription activity. This protein was also found to interact with helix-loop-helix protein E12 and is thought to be a negative regulator of liver-specific gene expression. Two alternatively spliced variants, which encode distinct isoforms, have been reported.Spezifikation
CCNDBP1 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O95273, Q3TVC7, Q5BK06 | |
17151, 23582, 362201 | |
CCNDBP1 Fusion Protein Ag3032 | |
Primary | |
-20°C | |
CCNDBP1 |
0.28 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CCNDBP1, Cyclin D1 binding protein 1, DIP1, GCIP, HHM, Human homolog of Maid | |
Ccndbp1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |