missing translation for 'onlineSavingsMsg'
Learn More

CCDC109B Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 20387-1-AP-20UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16839924

  • 133.80 CHF / 20 Mikroliter
Verfügbar ab: 23-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Negatively regulates the activity of MCU, the mitochondrial inner membrane calcium uniporter, and thereby modulates calcium uptake into the mitochondrion. Does not form functional calcium channels by itself. Mitochondrial calcium homeostasis plays key roles in cellular physiology and regulates cell bioenergetics, cytoplasmic calcium signals and activation of cell death pathways.
Spezifikation

Spezifikation

CCDC109B
Polyclonal
Unconjugated
MCUB
CCDC109B, MCUb
Rabbit
Antigen Affinity Chromatography
RUO
55013
-20°C
Liquid
Western Blot
0.28 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9NWR8
MCUB
CCDC109B Fusion Protein Ag14225
20 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote