missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase 3 Mouse anti-Human, Mouse, Rat, Clone: 2G4B2, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66470-2-IG-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This antibody can recognize p17, p19 and p32 of Caspase 3.
Smooth Muscle Actin belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actin being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. In particular, smooth muscle actin is an alpha actin that is found in skeletal muscle. Actin exists as a ubiquitous protein involved with filament formation that make up large portions of the cytoskeleton. Actin filaments interact with myosin to assist in muscle contraction as well as aiding in cell motility and cytokinesis. Smooth muscle actin is found on smooth muscle vessel walls, gut wall, myometrium, myoepithelial cells in breast and salivary glands. Defects in the smooth muscle actin gene cause aortic aneurysm familial thoracic type 6. Actin isoforms differ slightly in their N-terminus and the sequences of each are perfectly conserved in higher vertebrates.Spezifikation
Caspase 3 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P42574, P55213, P70677 | |
CASP3 | |
Caspase 3 Fusion Protein Ag25029 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
2G4B2 | |
Unconjugated | |
CASP3 | |
Apopain, CASP 3, CASP3, Caspase 3, Caspase3, Caspase-3, CPP 32, CPP32, CPP32B, Cysteine protease CPP32, Protein Yama, SCA 1, SREBP cleavage activity 1 | |
Mouse | |
Protein G | |
RUO | |
12367, 25402, 836 | |
-20°C | |
Liquid |