missing translation for 'onlineSavingsMsg'
Learn More

Caspase 3 Mouse anti-Human, Mouse, Rat, Clone: 2G4B2, Proteintech

Mouse Monoclonal Antibody

133.80 CHF - 381.34 CHF

Spezifikation

Antigen Caspase 3
Klon 2G4B2
Konzentration 1 mg/mL
Anwendungen Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Klassifikation Monoclonal
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
16848375
Unterlagen ansehen Aktionsdetails
Proteintech
66470-2-IG-20UL
20 μL
133.80 CHF
20 Mikroliter
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 17-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
16838375
Unterlagen ansehen Aktionsdetails
Proteintech
66470-2-IG-150UL
150 μL
381.34 CHF
150 Mikroliter
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 17-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

This antibody can recognize p17, p19 and p32 of Caspase 3.

Smooth Muscle Actin belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actin being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. In particular, smooth muscle actin is an alpha actin that is found in skeletal muscle. Actin exists as a ubiquitous protein involved with filament formation that make up large portions of the cytoskeleton. Actin filaments interact with myosin to assist in muscle contraction as well as aiding in cell motility and cytokinesis. Smooth muscle actin is found on smooth muscle vessel walls, gut wall, myometrium, myoepithelial cells in breast and salivary glands. Defects in the smooth muscle actin gene cause aortic aneurysm familial thoracic type 6. Actin isoforms differ slightly in their N-terminus and the sequences of each are perfectly conserved in higher vertebrates.
Spezifikation

Spezifikation

Caspase 3
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Apopain, CASP 3, CASP3, Caspase 3, Caspase3, Caspase-3, CPP 32, CPP32, CPP32B, Cysteine protease CPP32, Protein Yama, SCA 1, SREBP cleavage activity 1
CASP3
IgG1
Protein G
Antibody
2G4B2
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
Human, Mouse, Rat
P42574, P55213, P70677
12367, 25402, 836
Caspase 3 Fusion Protein Ag25029
Primary
-20°C
CASP3
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications