Learn More
Invitrogen™ Carbonic Anhydrase VA Monoclonal Antibody (K52035_18D12), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen™ MA556021
Beschreibung
Sequence of this protein is as follows: WQTSNNTLHP LWTVPVSVPG GTRQSPINIQ WRDSVYDPQL KPLRVSYEAA SCLYIWNTGY LFQVEFDDAT EASGISGGPL ENHYRLKQFH FHWGAVNEGG SEHTVDGHAY PAELHLVHWN SVKYQNYKEA VVGENGLAVI GVFLKLGAHH QTLQRLVDIL PEIKHKDARA AMRPFDPSTL LPTCWDYWTY AGSLTTPPLT ESVTWIIQKE PVEVAPSQLS AFRTLLFSAL GEEEKMMVNN YRPLQPLMNR KVWASFQATN
Carbonic anhydrases (CAs) are members of a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. CAs are involved in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric juice. They show extensive diversity in distribution and in their subcellular localization. The human CA5A gene encodes for the CA V protein which is expressed in the mitochondrial matrix in liver tissues, but can also be detected in many other tissues. In ureagenesis, the CA V protein may be involved in providing bicarbonate to carbamoyl phosphate synthetase.
Spezifikation
| Carbonic Anhydrase VA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| P35218, P43165 | |
| CA5A | |
| Recombinant human CA5A (immunogen range is 41-300). | |
| 200 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| K52035_18D12 | |
| Unconjugated | |
| CA5A | |
| CA Y; CA5; CA5A; CA5AD; Car5; Car5a; Carbonate dehydratase VA; carbonic anhydrase 5 mitochondrial; carbonic anhydrase 5, mitochondrial; carbonic anhydrase 5A; carbonic anhydrase 5a, mitochondrial; carbonic anhydrase 5b, mitochondrial; carbonic anhydrase V, mitochondrial; carbonic anhydrase VA; carbonic anhydrase VA, mitochondrial; carbonic dehydratase; CAV; CAVA; CA-VA; GS1-21A4.1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 54233, 763 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.