missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAMSAP2 Mouse anti-Human, Mouse, Rat, Clone: 2A10A1, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66463-1-IG-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
CAMSAP2 gene ontology annotations related to this gene include neuron projection development.Spezifikation
CAMSAP2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q08AD1, Q8C1B1 | |
Camsap2 | |
CAMSAP2 Fusion Protein Ag11794 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG1 |
Western Blot | |
2A10A1 | |
Unconjugated | |
CAMSAP2 | |
CAMSAP1L1, CAMSAP2, KIAA1078 | |
Mouse | |
Protein G | |
RUO | |
23271, 289400, 67886 | |
-20°C | |
Liquid |