missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CAMKK1 (CAMKKA) Recombinant Human Protein
CAMKK1 (CAMKKA) is a serine/threonine protein kinase family member.
Marke: Invitrogen™ PR8308A
Additional Details : Gewicht : 0.01000kg
Beschreibung
CAMKK1 (CAMKKA) belongs to the Ca2+/calmodulin-dependent protein kinase subfamily.
Spezifikation
NP_115670 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.02% Triton X-100, 0.5mM EDTA, 2mM DTT, 50 % Glycerol | |
100μg | |
RUO - research use only | |
CAMKK1 (CAMKKA) | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
59.8kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
CAMKK Kinase Family | |
Recombinant | |
Baculovirus |