missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C20orf149 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19912-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Specifications
C20orf149 | |
Polyclonal | |
Unconjugated | |
PPDPF | |
C20orf149, EXPDF, PPDPF, PPDPF,EXPDF | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
66496, 79144 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9CR37, Q9H3Y8 | |
Ppdpf | |
C20orf149 Fusion Protein Ag13740 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |