missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1orf77 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21456-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is a protein serine/threonine phosphatase of the PP2C family. This protein can interact with integrin-linked kinase (ILK/ILK1), a regulator of integrin mediated signaling, and regulate the kinase activity of ILK. Through the interaction with ILK, this protein may selectively affect the signaling process of ILK-mediated glycogen synthase kinase 3 beta (GSK3beta), and thus participate in Wnt signaling pathway.Spezifikation
C1orf77 | |
Polyclonal | |
Unconjugated | |
CHTOP | |
C1orf77, CHTOP, FOP, Friend of PRMT1 protein, pp7704, RP1 178F15.2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26097, 66511 | |
-20°C | |
Liquid |
Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9CY57, Q9Y3Y2 | |
Chtop | |
C1orf77 Fusion Protein Ag14736 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |