missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1orf77 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | C1orf77 |
---|---|
Konzentration | 0.19 mg/mL |
Anwendungen | Western Blot |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16851045
|
Proteintech
21456-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 09-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16841045
|
Proteintech
21456-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 09-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The protein encoded by this gene is a protein serine/threonine phosphatase of the PP2C family. This protein can interact with integrin-linked kinase (ILK/ILK1), a regulator of integrin mediated signaling, and regulate the kinase activity of ILK. Through the interaction with ILK, this protein may selectively affect the signaling process of ILK-mediated glycogen synthase kinase 3 beta (GSK3beta), and thus participate in Wnt signaling pathway.Spezifikation
C1orf77 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q9CY57, Q9Y3Y2 | |
26097, 66511 | |
C1orf77 Fusion Protein Ag14736 | |
Primary | |
-20°C | |
CHTOP |
0.19 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
C1orf77, CHTOP, FOP, Friend of PRMT1 protein, pp7704, RP1 178F15.2 | |
Chtop | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |