missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C11orf79 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 19906-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
19906-1-AP is a rabbit polyclonal antibody raised against the full-length of human SDHAF2.
SDHAF2, also named asC11orf79, SDHAF2 and SDH5, is a mitochondrial protein needed for the flavination of a succinate dehydrogenase complex subunit required for activity of the complex. SDHAF2 has been shown to be mutated in patients with paragangliomas, also known as familial non-chromaffin paragangliomas type 2.Spezifikation
C11orf79 | |
Polyclonal | |
Unconjugated | |
SDHAF2 | |
C11orf79, hSDH5, PGL2, SDH assembly factor 2, SDH5, SDHAF2, SDHAF2, PGL2, SDH5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
361726, 54949, 66072 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5RJQ7, Q8C6I2, Q9NX18 | |
Sdhaf2 | |
C11orf79 Fusion Protein Ag13893 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |