Learn More
BNP Antibody (50E1cc), Alexa Fluor™ 647, Novus Biologicals™
Artikelnummer. 18288324 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0.2 mL
Packungsgröße:
0.20 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18288324 0.2 mL 0.20 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 18288324 Lieferant Novus Biologicals Lieferanten-Nr. NB200440AF647
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Mouse Monoclonal Antibody
BNP Monoclonal antibody specifically detects BNP in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen BNP
Anwendungen Western Blot, ELISA
Klassifikation Monoclonal
Klon 50E1cc
Konjugat Alexa Fluor 647
Verdünnung Western Blot, ELISA, Sandwich ELISA
Zusammensetzung 50mM Sodium Borate
Gen-Alias BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide B, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein
Wirtsspezies Mouse
Immunogen Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with synthetic human BNP, whole molecule, conjugated with carrier protein.
Reinigungsverfahren Protein A purified
Menge 0.2 mL
Regulatorischer Status RUO
Forschungsgebiet Biologically Active Proteins, Cardiovascular Biology, Cytokine Research, Immunology, Neuroscience, Neurotransmission
Primär oder sekundär Primary
Gen-ID (Entrez) 4879
Zielspezies Human
Inhalt und Lagerung Store at 4°C in the dark.
Form Purified
Isotype IgG1