Learn More
BMSC UbP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11564-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
UBL7 (Ubiquitin Like 7) is a Protein Coding gene.Spezifikation
BMSC UbP | |
Polyclonal | |
Unconjugated | |
UBL7 | |
BMSC UbP, BMSCUBP, TCBA1, Ubiquitin like protein 7, Ubiquitin like protein SB132, UBL7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
300744, 69459, 84993 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q91W67, Q96S82 | |
UBL7 | |
BMSC UbP Fusion Protein Ag2130 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |