missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BBS1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21118-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Mutations in this gene have been observed in patients with the major form (type 1) of Bardet-Biedl syndrome. The encoded protein may play a role in eye, limb, cardiac and reproductive system development.Spezifikation
BBS1 | |
Polyclonal | |
Unconjugated | |
BBS1 | |
Bardet Biedl syndrome 1, BBS1, BBS2 like protein 2, BBS2L2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
52028, 582 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q3V3N7, Q8NFJ9 | |
Bbs1 | |
BBS1 Fusion Protein Ag15460 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |