missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BBS1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | BBS1 |
---|---|
Konzentration | 0.5 mg/mL |
Anwendungen | Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16850725
|
Proteintech
21118-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 08-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16840725
|
Proteintech
21118-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 08-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Mutations in this gene have been observed in patients with the major form (type 1) of Bardet-Biedl syndrome. The encoded protein may play a role in eye, limb, cardiac and reproductive system development.Spezifikation
BBS1 | |
Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
Q3V3N7, Q8NFJ9 | |
52028, 582 | |
BBS1 Fusion Protein Ag15460 | |
Primary | |
-20°C | |
BBS1 |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Bardet Biedl syndrome 1, BBS1, BBS2 like protein 2, BBS2L2 | |
Bbs1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |