missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASB9 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11952-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Substrate-recognition component of a SCF-like ECS (Elongin-Cullin-SOCS-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins. Recognizes at least two forms of creatine kinase, CKB and CKMT1A.Spezifikation
ASB9 | |
Polyclonal | |
Unconjugated | |
ASB9 | |
ASB 9, ASB9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
140462, 69299 | |
-20°C | |
Liquid |
Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q91ZT8, Q96DX5 | |
Asb9 | |
ASB9 Fusion Protein Ag2554 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |