missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASB9 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | ASB9 |
---|---|
Konzentration | 0.4 mg/mL |
Anwendungen | Western Blot |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16839363
|
Proteintech
11952-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 08-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16829363
|
Proteintech
11952-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 08-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Substrate-recognition component of a SCF-like ECS (Elongin-Cullin-SOCS-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins. Recognizes at least two forms of creatine kinase, CKB and CKMT1A.Spezifikation
ASB9 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q91ZT8, Q96DX5 | |
140462, 69299 | |
ASB9 Fusion Protein Ag2554 | |
Primary | |
-20°C | |
ASB9 |
0.4 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ASB 9, ASB9 | |
Asb9 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |