missing translation for 'onlineSavingsMsg'
Learn More

ARSK Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 19749-1-AP-20UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16839234

  • 133.80 CHF / 20 Mikroliter
Verfügbar ab: 17-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The antibody is specific to ARSK.

Sulfatases (EC 3.1.5.6), such as ARSK, hydrolyze sulfate esters from sulfated steroids, carbohydrates, proteoglycans, and glycolipids. They are involved in hormone biosynthesis, modulation of cell signaling, and degradation of macromolecules (Sardiello et al., 2005 ).
Spezifikation

Spezifikation

ARSK
Polyclonal
Unconjugated
ARSK
ARSK, arylsulfatase family, member K, Arylsulfatase K, ASK, Telethon sulfatase, TSULF
Rabbit
Antigen Affinity Chromatography
RUO
153642
-20°C
Liquid
Western Blot
0.58 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6UWY0
ARSK
Peptide
20 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote