missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARSK Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | ARSK |
---|---|
Konzentration | 0.58 mg/mL |
Anwendungen | Western Blot |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16839234
|
Proteintech
19749-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 07-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16829234
|
Proteintech
19749-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 07-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The antibody is specific to ARSK.
Sulfatases (EC 3.1.5.6), such as ARSK, hydrolyze sulfate esters from sulfated steroids, carbohydrates, proteoglycans, and glycolipids. They are involved in hormone biosynthesis, modulation of cell signaling, and degradation of macromolecules (Sardiello et al., 2005 ).Spezifikation
ARSK | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q6UWY0 | |
153642 | |
Peptide | |
Primary | |
-20°C | |
ARSK |
0.58 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ARSK, arylsulfatase family, member K, Arylsulfatase K, ASK, Telethon sulfatase, TSULF | |
ARSK | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |