missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARSK Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 19749-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The antibody is specific to ARSK.
Sulfatases (EC 3.1.5.6), such as ARSK, hydrolyze sulfate esters from sulfated steroids, carbohydrates, proteoglycans, and glycolipids. They are involved in hormone biosynthesis, modulation of cell signaling, and degradation of macromolecules (Sardiello et al., 2005 ).Spezifikation
ARSK | |
Polyclonal | |
Unconjugated | |
ARSK | |
ARSK, arylsulfatase family, member K, Arylsulfatase K, ASK, Telethon sulfatase, TSULF | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
153642 | |
-20°C | |
Liquid |
Western Blot | |
0.58 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6UWY0 | |
ARSK | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |