missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APOBEC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20121-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
APOBEC2 belongs to the cytidine and deoxycytidylate deaminase family. It is probable C to U editing enzyme whose physiological substrate is not yet known. It does not display detectable apoB mRNA editing and has a low intrinsic cytidine deaminase activity.Spezifikation
APOBEC2 | |
Polyclonal | |
Unconjugated | |
APOBEC2 | |
APOBEC2, ARCD1, ARP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10930, 11811, 301226 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.49 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9WV35, Q9Y235 | |
APOBEC2 | |
APOBEC2 Fusion Protein Ag13699 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |