missing translation for 'onlineSavingsMsg'
Learn More

APOBEC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 20121-1-AP-20UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16839614

  • 133.80 CHF / 20 Mikroliter
Verfügbar ab: 09-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

APOBEC2 belongs to the cytidine and deoxycytidylate deaminase family. It is probable C to U editing enzyme whose physiological substrate is not yet known. It does not display detectable apoB mRNA editing and has a low intrinsic cytidine deaminase activity.
Spezifikation

Spezifikation

APOBEC2
Polyclonal
Unconjugated
APOBEC2
APOBEC2, ARCD1, ARP1
Rabbit
Antigen Affinity Chromatography
RUO
10930, 11811, 301226
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot, Immunoprecipitation
0.49 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9WV35, Q9Y235
APOBEC2
APOBEC2 Fusion Protein Ag13699
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote