missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 19782-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Thymidylate kinase (DTYMK) catalyzes the phosphorylation of dTMP to form dTDP in the dTTP synthesis pathway for DNA synthesis. DTYMK is essential for DNA synthesis and is an important intermediate enzyme in the pathway of many pyrimidine analog drugs, including 5-Fluorouracil (5-FU), the chemotherapeutic drug of choice for several solid tumors. The human DTYMK gene was isolated via functional complementation of a Saccharomyces cerevisiae cell cycle mutant, cdc8. DTYMK mRNA levels and enzyme activities are regulated at the level of cell cycle and cell growth pathways.Spezifikation
APC | |
Polyclonal | |
Unconjugated | |
APC | |
adenomatous polyposis coli, APC, BTPS2, Deleted in polyposis 2.5, DP2, DP2.5, DP3, GS, Protein APC | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11789, 24205, 324 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P25054, P70478, Q61315 | |
APC | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |