Learn More
IBA Solutions For Life Science™ ApA-Dinukleotid-IBA
Artikelnummer. 12839376 Alle ansehen IBA Lifesciences Produkte
Ansicht ändern
Click to view available options
Menge:
1 μmol
Packungsgröße:
1 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
12839376 1 μmol 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 12839376 Lieferant IBA Solutions For Life Science™ Lieferanten-Nr. 50710164
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen
missing translation for 'validMainframeMsg'
Dinucleotides are used by RNA polymerases as initiators for RNA synthesis. Therefore, they are very useful for transcription analysis.
Spezifikation
Menge 1 μmol