missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP3M2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11925-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a subunit of the heterotetrameric adaptor-related protein complex 3 (AP-3), which belongs to the adaptor complexes medium subunits family. The AP-3 complex plays a role in protein trafficking to lysosomes and specialized organelles. Multiple alternatively spliced variants, encoding the same protein, have been identified.Spezifikation
AP3M2 | |
Polyclonal | |
Unconjugated | |
AP3M2 | |
AP 3 complex subunit mu 2, AP3M2, AP47B, CLA20, HA1 47 kDa subunit homolog 2, Mu3B adaptin, P47B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10947, 140667, 64933 | |
Antibody | |
IgG |
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P53677, P53678, Q8R2R9 | |
Ap3m2 | |
AP3M2 Fusion Protein Ag2572 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Liquid |