Learn More
RACK1 Mouse, Unlabeled, Clone: 20, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 610178
Additional Details : Gewicht : 0.01000kg
Beschreibung
Several proteins which specifically bind to PKC have been classified as RACKs (receptors for activated C-kinase). RACK1 was cloned from a rat brain cDNA expression library by screening for proteins that bind PKC in the presence of phosphatidylserine, diacylglycerol, and calcium in a PKC overlay assay. By sequence homology, RACK1 appears to belong to a superfamily that includes the β subunit of G proteins. All of these proteins contain five to eight internal repeat elements known as WD40 motifs, which appear to have a role in protein-protein interactions. In addition, RACK1 contains two short sequences homologous to a PKC-binding sequence identified in Annexin I and in the brain PKC inhibitor KCIP. The binding of RACK1 to PKC is dose-dependent and occurs at a site on PKC that is distinct from the catalytic domain, indicating that RACK1 is not a PKC substrate.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Spezifikation
RACK1 | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
150 μg | |
Cell Biology | |
Bovine, Canine, Chicken, Frog, Human, Mouse, Rat | |
IgM |
Western Blot | |
20 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.