Learn More
PP2A Catalytic α Mouse, Unlabeled, Clone: 46, BD
Mouse Monoclonal Antibody
370.00 CHF - 615.00 CHF
Spezifikation
Antigen | PP2A Catalytic α |
---|---|
Klon | 46 |
Konzentration | 250μg/mL |
Anwendungen | Western Blot |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15825519
|
BD Biosciences
610555 |
50 μg |
370.00 CHF
1 Set |
Verfügbar ab: 23-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15835519
|
BD Biosciences
610556 |
150 μg |
615.00 CHF
1 Set |
Verfügbar ab: 23-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
P rotein p hosphatases of the type 2A (PP2A) are composed of two regulatory and one catalytic subunit. The different subunits are encoded by multiple genes. PP2A are specific for phosphoserine and phosphothreonine residues of proteins that are involved in the regulation of cellular growth. PP2A binds and dephosphorylates the middle T antigens of polyoma virus. This interaction may regulate the transcriptional activity of the virus. The 36 kDa catalytic subunit is rapidly tyrosine phosphorylated as a result of growth factor stimulation or cell transformation. Tyrosine phosphorylation of the catalytic subunit may inactivate the phosphatase activity of PP2A.
Spezifikation
PP2A Catalytic α | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
46 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Human, Mouse, Rat | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.