missing translation for 'onlineSavingsMsg'
Learn More

PP2A Catalytic α Mouse, Unlabeled, Clone: 46, BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 610556

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15835519

  • 615.00 CHF / 1 Set
Verfügbar ab: 17-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

P rotein p hosphatases of the type 2A (PP2A) are composed of two regulatory and one catalytic subunit. The different subunits are encoded by multiple genes. PP2A are specific for phosphoserine and phosphothreonine residues of proteins that are involved in the regulation of cellular growth. PP2A binds and dephosphorylates the middle T antigens of polyoma virus. This interaction may regulate the transcriptional activity of the virus. The 36 kDa catalytic subunit is rapidly tyrosine phosphorylated as a result of growth factor stimulation or cell transformation. Tyrosine phosphorylation of the catalytic subunit may inactivate the phosphatase activity of PP2A.

Spezifikation

Spezifikation

PP2A Catalytic α
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
150 μg
Cell Biology
Canine, Human, Mouse, Rat
IgG1
Western Blot
46
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only.