Learn More
anti-IgM κ, Mouse, FITC, Clone: G155-228, Isotype Control, BD
Marke: BD Biosciences 553474
Additional Details : Gewicht : 0.01000kg
Beschreibung
The G155-228 antibody is specific for the hapten trinitrophenol (TNP), a hapten not expressed on non-human primate cells or non-human primate cell lines. In the absence of specific binding, this immunoglobulin may bind non-specifically to Fc receptors.
Host Species: Mouse
Clone: G155-228
Isotype: IgM, κ
Species Reactivity: Non-human primate
Immunogen: TNP-Keyhole Limpet Hemocyanin
Flow Cytometry, Immunofluorescence, Intracellular Staining
Spezifikation
Flow Cytometry, Immunofluorescence, Western Blot | |
0.5mg/mL | |
Mouse | |
Affinity Purified | |
Non-human Primate | |
IgM κ |
G155-228 | |
FITC | |
TNP-keyhole limpet hemocyanin | |
0.25 mg | |
Undiluted at 4°C. Protect from exposure to light. Do not freeze. |